Product Details

Assay Reference Genome
Location

Chr.15:30972922 on build GRCm38
Cytoband
15B2
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
18163
Gene Symbol
Ctnnd2
Gene Name
catenin (cadherin associated protein), delta 2
Gene Aliases
Catnd2, Nprap, neurojugin
Location
Chr.15:30172593-31029344 on build GRCm38
Assay Gene Location
Overlaps Exon 19 - Intron 19
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Ctnnd2 NM_008729.2 NP_032755.2
AK140454.1 BAE24395.1
AK140960.1 BAE24530.1
AK164304.1 BAE37727.1
BC138260.1
BC138261.1
BC145217.1
U90331.1 AAB82409.1

View Full Product Details