Product Details
- Assay Reference Genome
Location
Chr.15:30887272 on build GRCm38
- Cytoband
- 15B2
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 18163
- Gene Symbol
- Ctnnd2
- Gene Name
- catenin (cadherin associated protein), delta 2
- Gene Aliases
- Catnd2, Nprap, neurojugin
- Location
- Chr.15:30172593-31029344 on build GRCm38
- Assay Gene Location
- Overlaps Exon 14 - Intron 14
View Full Product Details