Product Details
- Assay Reference Genome
Location
Chr.15:47676471 on build GRCm38
- Cytoband
- 15B3.3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 239420
- Gene Symbol
- Csmd3
- Gene Name
- CUB and Sushi multiple domains 3
- Gene Aliases
- 4930500N14Rik, mKIAA1894
- Location
- Chr.15:47580633-48792231 on build GRCm38
- Assay Gene Location
- Overlaps Intron 45 - Exon 46
View Full Product Details