Product Details
- Assay Reference Genome
Location
Chr.14:28943697 on build GRCm38
- Cytoband
- 14A3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 12294
- Gene Symbol
- Cacna2d3
- Gene Name
- calcium channel, voltage-dependent, alpha2/delta subunit 3
- Gene Aliases
- Cacnad3
- Location
- Chr.14:28904943-29721864 on build GRCm38
- Assay Gene Location
- Overlaps Intron 34 - Exon 35
View Full Product Details