Product Details

Assay Reference Genome
Location

Chr.12:88954823 on build GRCm38
Cytoband
12D3
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
18191
Gene Symbol
Nrxn3
Gene Name
neurexin III
Gene Aliases
Location
Chr.12:88722865-90334935 on build GRCm38
Assay Gene Location
Overlaps Intron 4 - Exon 5
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Nrxn3 NM_001198587.3 NP_001185516.2
NM_172544.3 NP_766132.2
AK032126.1 BAC27716.1
AK078216.1
AK082055.1
AK129205.1 BAC98015.2
BC060719.1 AAH60719.1
CJ104960.1

View Full Product Details