Product Details
- Assay Reference Genome
Location
Chr.12:78626162 on build GRCm38
- Cytoband
- 12C3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 268566
- Gene Symbol
- Gphn
- Gene Name
- gephyrin
- Gene Aliases
- 5730552E08Rik, AI662856, BC027112, C230040D23, GPH, GPHRYN, geph
- Location
- Chr.12:78224153-78684772 on build GRCm38
- Assay Gene Location
- Overlaps Intron 16 - Intron 17
View Full Product Details