Product Details
- Assay Reference Genome
Location
Chr.12:54062021 on build GRCm38
- Cytoband
- 12C1
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 27386
- Gene Symbol
- Npas3
- Gene Name
- neuronal PAS domain protein 3
- Gene Aliases
- 4930423H22Rik, bHLHe12
- Location
- Chr.12:53248003-54072175 on build GRCm38
- Assay Gene Location
- Overlaps Intron 9 - Exon 10
View Full Product Details