Product Details
- Assay Reference Genome
Location
Chr.11:36049084 on build GRCm38
- Cytoband
- 11A4
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 23964
- Gene Symbol
- Tenm2
- Gene Name
- teneurin transmembrane protein 2
- Gene Aliases
- Odz2, Odz3, Ten-2, Ten-m2
- Location
- Chr.11:36006656-37236808 on build GRCm38
- Assay Gene Location
- Overlaps Intron 22 - Exon 22
View Full Product Details