Product Details
- Assay Reference Genome
Location
Chr.11:34401054 on build GRCm38
- Cytoband
- 11A4
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 574403
- Gene Symbol
- Fam196b
- Gene Name
- family with sequence similarity 196, member B
- Gene Aliases
- EG574403, Gm6041
- Location
- Chr.11:34314822-34422642 on build GRCm38
- Assay Gene Location
- Overlaps Intron 1 - Exon 2
View Full Product Details