Product Details
- Assay Reference Genome
Location
Chr.5:131533926 on build GRCm38
- Cytoband
- 5G2
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 319974
- Gene Symbol
- Auts2
- Gene Name
- autism susceptibility candidate 2
- Gene Aliases
- 2700063G02Rik, A730011F23Rik, D830032G16Rik
- Location
- Chr.5:131437682-132542343 on build GRCm38
- Assay Gene Location
- Within Intron 5
View Full Product Details