Product Details

Assay Reference Genome
Location

Chr.2:161698076 on build GRCm38
Cytoband
2H2
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
19281
Gene Symbol
Ptprt
Gene Name
protein tyrosine phosphatase, receptor type, T
Gene Aliases
R-PTP-T, RPTP-rfo, RPTP-rho, RPTPmam4, RPTPrho, mKIAA0283, mRPTPrho
Location
Chr.2:161521988-162661147 on build GRCm38
Assay Gene Location
Overlaps Intron 13 - Exon 14
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Ptprt NM_001291149.1 NP_001278078.1
NM_001291150.1 NP_001278079.1
NM_001291151.1 NP_001278080.1
NM_021464.5 NP_067439.1
AF152556.4 AAD34158.4
AF162856.2 AAF82400.2
AF162857.1 AAF82401.1
AF244125.1 AAF44712.1
AK220254.1 BAD90179.1
AY026861.1 AAK18741.1
AY026862.1 AAK18742.1
AY026863.1 AAK18743.1

View Full Product Details