Product Details
- Assay Reference Genome
Location
Chr.2:142176549 on build GRCm38
- Cytoband
- 2G1
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 72899
- Gene Symbol
- Macrod2
- Gene Name
- MACRO domain containing 2
- Gene Aliases
- 1110033L15Rik, 2610107G07Rik, 2900006F19Rik
- Location
- Chr.2:140395356-142392966 on build GRCm38
- Assay Gene Location
- Overlaps Intron 10 - Exon 11
View Full Product Details