Product Details
- Assay Reference Genome
Location
Chr.2:40701771 on build GRCm38
- Cytoband
- 2B
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 94217
- Gene Symbol
- Lrp1b
- Gene Name
- low density lipoprotein-related protein 1B (deleted in tumors)
- Gene Aliases
- 9630004P12Rik, LRP-DIT
- Location
- Chr.2:40595290-42653631 on build GRCm38
- Assay Gene Location
- Overlaps Intron 75 - Exon 75
View Full Product Details