Product Details

Assay Reference Genome
Location

Chr.1:69827092 on build GRCm38
Cytoband
1C3
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
66722
Gene Symbol
Spag16
Gene Name
sperm associated antigen 16
Gene Aliases
4921511D23Rik, 4930524F24Rik, 4930585K05Rik, AV261009, Pf20, Wdr29
Location
Chr.1:69826872-70725132 on build GRCm38
Assay Gene Location
Overlaps Exon 1 - Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Spag16 NM_001271533.1 NP_001258462.1
NM_029160.3 NP_083436.2
AF490391.1 AAM97148.1
AK015884.1 BAB30016.1
AK083083.1 BAC38755.1
BC120668.1
BC145006.1

View Full Product Details