Product Details
- Assay Reference Genome
Location
Chr.1:62238972 on build GRCm38
- Cytoband
- 1C2
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 72823
- Gene Symbol
- Pard3b
- Gene Name
- par-3 family cell polarity regulator beta
- Gene Aliases
- 1810008K04Rik, 2010002N16Rik, 2810455B10Rik, Als2cr19, PAR3B, PAR3L, PAR3beta
- Location
- Chr.1:61638667-62642284 on build GRCm38
- Assay Gene Location
- Overlaps Exon 15 - Intron 15
View Full Product Details