Product Details

Assay Reference Genome
Location

Chr.1:25074655 on build GRCm38
Cytoband
1A5
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
210933
Gene Symbol
Adgrb3
Gene Name
adhesion G protein-coupled receptor B3
Gene Aliases
A830096D10Rik, Bai3
Location
Chr.1:25067476-25830206 on build GRCm38
Assay Gene Location
Overlaps Intron 30 - Exon 30
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Adgrb3 NM_175642.4 NP_783573.4
AK044248.1 BAC31839.1
AK081421.1 BAC38215.1
AY168406.1 AAO27431.1
BC032251.1 AAH32251.1
BC079670.1 AAH79670.1
BC099951.1 AAH99951.1

View Full Product Details