Product Details
- Assay Reference Genome
Location
Chr.5:169865249 on build GRCh38
- Cytoband
- 5q35.1
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 1794
- Gene Symbol
- DOCK2
- Gene Name
- dedicator of cytokinesis 2
- Gene Aliases
- IMD40
- Location
- Chr.5:169637247-170083382 on build GRCh38
- Assay Gene Location
- Within Intron 29
- Entrez Gene ID
- 100131897
- Gene Symbol
- FAM196B
- Gene Name
- family with sequence similarity 196 member B
- Gene Aliases
- C5orf57
- Location
- Chr.5:169861304-169980740 on build GRCh38
- Assay Gene Location
- Within Intron 3
View Full Product Details