Product Details
- Assay Reference Genome
Location
Chr.14:32953367 on build GRCh38
- Cytoband
- 14q13.1
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 64067
- Gene Symbol
- NPAS3
- Gene Name
- neuronal PAS domain protein 3
- Gene Aliases
- MOP6, PASD6, bHLHe12
- Location
- Chr.14:32934785-33804176 on build GRCh38
- Assay Gene Location
- Within Intron 2
View Full Product Details