Product Details

Assay Reference Genome
Location

Chr.13:93246008 on build GRCh38
Cytoband
13q31.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
10082
Gene Symbol
GPC6
Gene Name
glypican 6
Gene Aliases
OMIMD1
Location
Chr.13:93226825-94408020 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GPC6 NM_005708.3 NP_005699.1
AF105267.1 AAD55749.1
AF111178.1 AAD31392.1
AK074741.1
AK130013.1
AK290144.1
AK304338.1
AY358462.1 AAQ88827.1
BC106947.2 AAI06948.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3582694 Chr13:91877905 - 93587318 on Build GRCh38 Gain GPC6 GPC5-AS1 GPC5 GPC5-AS2 MIR548AS LINC00363
esv3892424 Chr13:92937463 - 94196357 on Build GRCh38 Gain GPC6-AS2 GPC6-AS1 GPC6 LINC00363

View Full Product Details