Product Details
- Assay Reference Genome
Location
Chr.3:173406729 on build GRCh38
- Cytoband
- 3q26.31
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 22871
- Gene Symbol
- NLGN1
- Gene Name
- neuroligin 1
- Gene Aliases
- NL1
- Location
- Chr.3:173395899-174291287 on build GRCh38
- Assay Gene Location
- Within Intron 2
View Full Product Details