Product Details

Assay Reference Genome
Location

Chr.1:57014093 on build GRCh38
Cytoband
1p32.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1600
Gene Symbol
DAB1
Gene Name
DAB1, reelin adaptor protein
Gene Aliases
Location
Chr.1:56997906-58250539 on build GRCh38
Assay Gene Location
Within Intron 14
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
DAB1 NM_021080.3 NP_066566.3
AB210012.1
AF071062.1 AAC70068.1
AK095513.1
AK313144.1
BC067445.1 AAH67445.1
BC067446.1 AAH67446.1
BC067447.1 AAH67447.1
BC112938.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv546305 Chr1:56997653 - 57350817 on Build GRCh38 Gain LOC105378749 DAB1

View Full Product Details