Product Details

Assay Reference Genome
Location

Chr.1:13928762 on build GRCh38
Cytoband
1p36.21
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
23254
Gene Symbol
KAZN
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
KAZ
Location
Chr.1:13893387-15118048 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
KAZN XM_005245795.4 XP_005245852.1
XM_011541074.2 XP_011539376.1
XM_011541080.2 XP_011539382.1
XM_017000768.1 XP_016856257.1
XM_017000769.1 XP_016856258.1
XM_017000770.1 XP_016856259.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv545567 Chr1:13558784 - 14026576 on Build GRCh38 Gain PRDM2 PDPN KAZN

View Full Product Details