Product Details

Assay Reference Genome
Location

Chr.18:52362284 on build GRCh38
Cytoband
18q21.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1630
Gene Symbol
DCC
Gene Name
DCC netrin 1 receptor
Gene Aliases
CRC18, CRCR1, IGDCC1, MRMV1, NTN1R1
Location
Chr.18:52340172-53535903 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
DCC NM_005215.3 NP_005206.2
XM_011525843.1 XP_011524145.1
XM_017025568.1 XP_016881057.1
XM_017025569.1 XP_016881058.1
M32292.1 AAA35751.1
X76132.1 CAA53735.1

View Full Product Details