Product Details

Assay Reference Genome
Location

Chr.13:91400316 on build GRCh38
Cytoband
13q31.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2262
Gene Symbol
GPC5
Gene Name
glypican 5
Gene Aliases
Location
Chr.13:91398619-92867237 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GPC5 NM_004466.5 NP_004457.1
XM_011521054.2 XP_011519356.1
XM_011521055.2 XP_011519357.1
XM_011521056.2 XP_011519358.1
XM_011521057.2 XP_011519359.1
XM_011521058.2 XP_011519360.1
XM_011521059.2 XP_011519361.1
XM_011521060.2 XP_011519362.1
XM_017020435.1 XP_016875924.1
XM_017020436.1 XP_016875925.1
XM_017020437.1 XP_016875926.1
AF001462.1 AAC12261.1
AK312815.1
BC039730.1 AAH39730.1
DB100549.1
U66033.1 AAC51118.1

View Full Product Details