Product Details
- Assay Reference Genome
Location
Chr.12:23539589 on build GRCh38
- Cytoband
- 12p12.1
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 6660
- Gene Symbol
- SOX5
- Gene Name
- SRY-box 5
- Gene Aliases
- L-SOX5, L-SOX5B, L-SOX5F, LAMSHF
- Location
- Chr.12:23529499-24562701 on build GRCh38
- Assay Gene Location
- Within Intron 24
View Full Product Details