Product Details

Assay Reference Genome
Location

Chr.11:131392971 on build GRCh38
Cytoband
11q25
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
50863
Gene Symbol
NTM
Gene Name
neurotrimin
Gene Aliases
HNT, IGLON2, NTRI
Location
Chr.11:131370326-132336822 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NTM NM_001048209.1 NP_001041674.1
XM_005271580.3 XP_005271637.1
XM_005271582.3 XP_005271639.1
XM_006718855.3 XP_006718918.1
XM_017017852.1 XP_016873341.1
XM_017017853.1 XP_016873342.1
XM_017017855.1 XP_016873344.1
XM_017017861.1 XP_016873350.1
XM_017017865.1 XP_016873354.1
AK293487.1
AK294137.1
AK315964.1
AY358331.1 AAQ88697.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3892109 Chr11:131358314 - 131494601 on Build GRCh38 Loss NTM
nsv1035733 Chr11:130509478 - 131429821 on Build GRCh38 Gain LOC103611081 C11orf44 SNX19 NTM LOC100507431 MIR8052
nsv1039087 Chr11:131230668 - 131438530 on Build GRCh38 Gain NTM

View Full Product Details