Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation ID |
Location |
CNV Subtype |
Genes |
dgv65n21 |
Chr11:40049473 - 40249892 on Build GRCh38 |
Gain |
LRRC4C
|
Product Details
- Assay Reference Genome
Location
Chr.11:40135942 on build GRCh38
- Cytoband
- 11p12
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 57689
- Gene Symbol
- LRRC4C
- Gene Name
- leucine rich repeat containing 4C
- Gene Aliases
- NGL-1, NGL1
- Location
- Chr.11:40107066-41460212 on build GRCh38
- Assay Gene Location
- Within Intron 9
Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation |
Location |
CNV Subtype |
Genes |
dgv65n21 |
Chr11:40049473 - 40249892 on Build GRCh38 |
Gain |
LRRC4C
|
View Full Product Details