Product Details
- Assay Reference Genome
Location
Chr.11:19349809 on build GRCh38
- Cytoband
- 11p15.1
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 89797
- Gene Symbol
- NAV2
- Gene Name
- neuron navigator 2
- Gene Aliases
- HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
- Location
- Chr.11:19345200-20121601 on build GRCh38
- Assay Gene Location
- Within Intron 1
View Full Product Details