Product Details

Assay Reference Genome
Location

Chr.11:19362388 on build GRCh38
Cytoband
11p15.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
89797
Gene Symbol
NAV2
Gene Name
neuron navigator 2
Gene Aliases
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Location
Chr.11:19345200-20121601 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NAV2 NM_001111018.1 NP_001104488.1
XM_011520452.2 XP_011518754.1
XM_017018520.1 XP_016874009.1
XM_017018522.1 XP_016874011.1
AB063115.1 BAC00853.1

View Full Product Details