Product Details

Assay Reference Genome
Location

Chr.8:31667226 on build GRCh38
Cytoband
8p12
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
3084
Gene Symbol
NRG1
Gene Name
neuregulin 1
Gene Aliases
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, NRG1-IT2, SMDF
Location
Chr.8:31639222-32771716 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NRG1 NM_001159995.2 NP_001153467.1
NM_001159999.2 NP_001153471.1
NM_001160001.2 NP_001153473.1
NM_001322201.1 NP_001309130.1
NM_001322202.1 NP_001309131.1
NM_013962.2 NP_039256.2
XM_011544512.2 XP_011542814.1
XM_017013365.1 XP_016868854.1
XM_017013366.1 XP_016868855.1
XM_017013367.1 XP_016868856.1
XM_017013371.1 XP_016868860.1
XM_017013372.1 XP_016868861.1
CN603652.1
CN603654.1
EF372273.1
EF372274.1
EF372275.1
EF372276.1
EF372277.1
EF517295.1
EF517296.1
EF517297.1
GQ983557.1
GQ983558.1
L12260.1 AAB59622.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3891380 Chr8:31278415 - 31706814 on Build GRCh38 Gain NRG1

View Full Product Details