Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation ID |
Location |
CNV Subtype |
Genes |
esv3891380 |
Chr8:31278415 - 31706814 on Build GRCh38 |
Gain |
NRG1
|
Product Details
- Assay Reference Genome
Location
Chr.8:31650670 on build GRCh38
- Cytoband
- 8p12
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 3084
- Gene Symbol
- NRG1
- Gene Name
- neuregulin 1
- Gene Aliases
- ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, NRG1-IT2, SMDF
- Location
- Chr.8:31639222-32771716 on build GRCh38
- Assay Gene Location
- Within Intron 2
Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation |
Location |
CNV Subtype |
Genes |
esv3891380 |
Chr8:31278415 - 31706814 on Build GRCh38 |
Gain |
NRG1
|
View Full Product Details