Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation ID |
Location |
CNV Subtype |
Genes |
dgv5823n100 |
Chr5:155731480 - 156135824 on Build GRCh38 |
Gain |
SGCD
|
Product Details
- Assay Reference Genome
Location
Chr.5:155747897 on build GRCh38
- Cytoband
- 5q33.2
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 6444
- Gene Symbol
- SGCD
- Gene Name
- sarcoglycan delta
- Gene Aliases
- 35DAG, CMD1L, DAGD, SG-delta, SGCDP, SGD
- Location
- Chr.5:155728636-156767788 on build GRCh38
- Assay Gene Location
- Within Intron 1
Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation |
Location |
CNV Subtype |
Genes |
dgv5823n100 |
Chr5:155731480 - 156135824 on Build GRCh38 |
Gain |
SGCD
|
View Full Product Details