Product Details

Assay Reference Genome
Location

Chr.5:155739681 on build GRCh38
Cytoband
5q33.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
6444
Gene Symbol
SGCD
Gene Name
sarcoglycan delta
Gene Aliases
35DAG, CMD1L, DAGD, SG-delta, SGCDP, SGD
Location
Chr.5:155728636-156767788 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SGCD XM_017009724.1 XP_016865213.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3607264 Chr5:155733910 - 155740456 on Build GRCh38 Loss SGCD
dgv5823n100 Chr5:155731480 - 156135824 on Build GRCh38 Gain SGCD

View Full Product Details