Product Details

Assay Reference Genome
Location

Chr.5:19478668 on build GRCh38
Cytoband
5p14.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1016
Gene Symbol
CDH18
Gene Name
cadherin 18
Gene Aliases
CDH14, CDH14L, CDH24
Location
Chr.5:19472930-20575902 on build GRCh38
Assay Gene Location
Within Intron 23
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CDH18 NM_001167667.1 NP_001161139.1
NM_001291956.1 NP_001278885.1
NM_001291957.1 NP_001278886.1
NM_004934.3 NP_004925.1
XM_005248228.3 XP_005248285.1
XM_006714435.3 XP_006714498.1
XM_011513928.2 XP_011512230.1
XM_011513929.2 XP_011512231.1
XM_011513930.2 XP_011512232.1
XM_017008924.1 XP_016864413.1
XM_017008925.1 XP_016864414.1
XM_017008926.1 XP_016864415.1
XM_017008927.1 XP_016864416.1
XM_017008928.1 XP_016864417.1
XM_017008929.1 XP_016864418.1
XM_017008930.1 XP_016864419.1
XM_017008931.1 XP_016864420.1
XM_017008932.1 XP_016864421.1
XM_017008933.1 XP_016864422.1
XM_017008934.1 XP_016864423.1
XM_017008935.1 XP_016864424.1
XM_017008936.1 XP_016864425.1
XM_017008937.1 XP_016864426.1
XM_017008938.1 XP_016864427.1
XM_017008939.1 XP_016864428.1
XM_017008940.1 XP_016864429.1
XM_017008941.1 XP_016864430.1
AK289547.1
AK295087.1
AK308745.1
BC031051.1 AAH31051.1
U59325.1 AAB02933.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1031377 Chr5:19289979 - 20971814 on Build GRCh38 Loss CDH18 LOC105374676
nsv597424 Chr5:19381946 - 19751051 on Build GRCh38 Gain CDH18
nsv470997 Chr5:19441626 - 19534696 on Build GRCh38 Loss CDH18
nsv4752 Chr5:19449761 - 19482880 on Build GRCh38 Insertion CDH18

View Full Product Details