Product Details

Assay Reference Genome
Location

Chr.5:169640318 on build GRCh38
Cytoband
5q35.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1794
Gene Symbol
DOCK2
Gene Name
dedicator of cytokinesis 2
Gene Aliases
IMD40
Location
Chr.5:169637247-170083382 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
DOCK2 NM_004946.2 NP_004937.1
XM_005265830.3 XP_005265887.1
XM_011534448.2 XP_011532750.1
XM_011534449.2 XP_011532751.1
XM_011534450.2 XP_011532752.1
XM_011534451.2 XP_011532753.1
XM_017009189.1 XP_016864678.1
XM_017009190.1 XP_016864679.1
BC104900.1 AAI04901.1
BC113457.1
D86964.1
DB145764.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv5124 Chr5:169631419 - 169676697 on Build GRCh38 Deletion DOCK2
nsv1031257 Chr5:169272339 - 169832306 on Build GRCh38 Gain SPDL1 SLIT3 DOCK2

View Full Product Details