Product Details

Assay Reference Genome
Location

Chr.4:92318384 on build GRCh38
Cytoband
4q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2895
Gene Symbol
GRID2
Gene Name
glutamate ionotropic receptor delta type subunit 2
Gene Aliases
GluD2, SCAR18
Location
Chr.4:92304249-93810330 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GRID2 NM_001286838.1 NP_001273767.1
NM_001510.3 NP_001501.2
XM_017008122.1 XP_016863611.1
AB209318.1 BAD92555.1
AF009014.1 AAC39579.1
AK308611.1
BC099652.3 AAH99652.1
BC099653.3 AAH99653.1
BC099654.3 AAH99654.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv594868 Chr4:92269047 - 92470100 on Build GRCh38 Gain LOC101929194 GRID2
nsv830007 Chr4:92180761 - 92396505 on Build GRCh38 Gain LOC101929194 GRID2
nsv1003867 Chr4:91799329 - 93620435 on Build GRCh38 Gain LOC101929194 GRID2
esv3893983 Chr4:90539430 - 92587744 on Build GRCh38 Gain LOC101929194 CCSER1 GRID2
nsv998246 Chr4:92277694 - 92429174 on Build GRCh38 Gain GRID2
esv3601396 Chr4:92306604 - 92347058 on Build GRCh38 Gain GRID2
esv3601393 Chr4:92178641 - 92502792 on Build GRCh38 Gain LOC101929194 GRID2
nsv1009960 Chr4:91826380 - 92467903 on Build GRCh38 Gain LOC101929194 GRID2
nsv594866 Chr4:91916433 - 92455080 on Build GRCh38 Gain LOC101929194 GRID2

View Full Product Details