Product Details

Assay Reference Genome
Location

Chr.4:20754492 on build GRCh38
Cytoband
4p15.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
80333
Gene Symbol
KCNIP4
Gene Name
potassium voltage-gated channel interacting protein 4
Gene Aliases
CALP, KCHIP4
Location
Chr.4:20728520-21948801 on build GRCh38
Assay Gene Location
Within Intron 10
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
KCNIP4 NM_001035003.1 NP_001030175.1
NM_001035004.1 NP_001030176.1
NM_025221.5 NP_079497.2
NM_147181.3 NP_671710.1
NM_147182.3 NP_671711.1
NM_147183.3 NP_671712.1
XM_005248190.4 XP_005248247.1
XM_011513882.2 XP_011512184.1
XM_011513883.2 XP_011512185.1
XM_011513884.2 XP_011512186.1
XM_011513885.2 XP_011512187.1
XM_011513886.2 XP_011512188.1
XM_011513887.2 XP_011512189.1
XM_011513889.2 XP_011512191.1
XM_017008653.1 XP_016864142.1
AF302044.1 AAG36974.1
AF305072.1 AAG36977.1
AF367023.1 AAK53712.1
AF367024.1 AAK53713.1
AF453246.1 AAL86769.1
AK289661.1
AK289922.1
AK290998.1
AK294188.1
AY029176.1 AAK31594.1
AY118170.1 AAM81578.1
BC032520.1 AAH32520.1
BX648990.1
DQ148487.1 AAZ77804.1
DQ148488.1 AAZ77805.1
DQ148489.1 AAZ77806.1
DQ148490.1 AAZ77807.1
DQ148491.1 AAZ77808.1
DQ148492.1 AAZ77809.1
Entrez Gene ID
133015
Gene Symbol
PACRGL
Gene Name
PARK2 coregulated like
Gene Aliases
C4orf28
Location
Chr.4:20696282-20757766 on build GRCh38
Assay Gene Location
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PACRGL

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv829876 Chr4:20637113 - 20833998 on Build GRCh38 Gain KCNIP4 PACRGL

View Full Product Details