Product Details

Assay Reference Genome
Location

Chr.4:171834094 on build GRCh38
Cytoband
4q34.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
442117
Gene Symbol
GALNTL6
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 6
Gene Aliases
GALNACT20, GALNT17, GalNAc-T6L
Location
Chr.4:171812690-173040408 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GALNTL6 NM_001034845.2 NP_001030017.2
XM_017008243.1 XP_016863732.1
AK295464.1
BC047551.1 AAH47551.1
GU220060.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv461838 Chr4:171340402 - 172518114 on Build GRCh38 Gain GALNTL6 LOC441052
nsv596171 Chr4:171184677 - 172359870 on Build GRCh38 Loss MIR6082 LOC101928288 GALNTL6 LOC441052

View Full Product Details