Product Details

Assay Reference Genome
Location

Chr.3:6880751 on build GRCh38
Cytoband
3p26.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2917
Gene Symbol
GRM7
Gene Name
glutamate metabotropic receptor 7
Gene Aliases
GLUR7, GPRC1G, MGLU7, MGLUR7, PPP1R87
Location
Chr.3:6861097-7743032 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GRM7 NM_000844.3 NP_000835.1
NM_181874.2 NP_870989.1
XM_017006272.1 XP_016861761.1
XM_017006273.1 XP_016861762.1
AK312488.1
BC136458.1
BC136459.1
X94552.1 CAA64245.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv3893654 Chr3:6811906 - 7026460 on Build GRCh38 Loss GRM7 GRM7-AS2
esv2763214 Chr3:6753213 - 7218258 on Build GRCh38 Gain GRM7-AS3 GRM7 GRM7-AS2
nsv518755 Chr3:6878131 - 6942063 on Build GRCh38 Loss GRM7 GRM7-AS2
nsv1006252 Chr3:6707783 - 7544588 on Build GRCh38 Gain GRM7-AS3 GRM7-AS1 GRM7 LOC105376944 GRM7-AS2
nsv589568 Chr3:6713211 - 7549718 on Build GRCh38 Gain GRM7-AS3 GRM7-AS1 GRM7 LOC105376944 GRM7-AS2
dgv4674n100 Chr3:6680357 - 7004828 on Build GRCh38 Gain GRM7-AS3 GRM7 LOC105376944 GRM7-AS2

View Full Product Details