Product Details

Assay Reference Genome
Location

Chr.2:213294673 on build GRCh38
Cytoband
2q34
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
79582
Gene Symbol
SPAG16
Gene Name
sperm associated antigen 16
Gene Aliases
PF20, WDR29
Location
Chr.2:213284369-214410501 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SPAG16 NM_001025436.2 NP_001020607.1
NM_024532.4 NP_078808.3
NR_047659.1
NR_047660.1
XM_006712746.2 XP_006712809.1
XM_011511814.1 XP_011510116.1
XM_011511815.1 XP_011510117.1
XM_011511816.2 XP_011510118.1
XM_011511817.1 XP_011510119.1
XM_011511818.1 XP_011510120.1
XM_011511820.1 XP_011510122.1
XM_011511821.2 XP_011510123.1
XM_011511822.1 XP_011510124.1
XM_011511824.1 XP_011510126.1
XM_011511827.1 XP_011510129.1
XM_011511828.1 XP_011510130.1
XM_011511829.1 XP_011510131.1
XM_011511831.1 XP_011510133.1
XM_011511832.1 XP_011510134.1
XM_011511835.2 XP_011510137.1
XM_011511837.2 XP_011510139.1
XM_017004896.1 XP_016860385.1
XM_017004899.1 XP_016860388.1
XM_017004900.1 XP_016860389.1
XM_017004901.1 XP_016860390.1
AF310672.1 AAM63956.1
AF426740.1 AAN63530.1
AF490390.1 AAM97147.1
AK026377.1 BAB15466.1
AK095036.1 BAC04481.1
AK302351.1
AK310883.1
AL832962.1 CAH56274.1
BC009614.1 AAH09614.1
BC025379.1 AAH25379.1
BC067756.1 AAH67756.1
BC100282.1 AAI00283.1
CR457359.1 CAG33640.1
CR749477.1 CAH18307.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv3152 Chr2:213282926 - 213332692 on Build GRCh38 Insertion LOC100130451 SPAG16
nsv1013206 Chr2:213267855 - 213302172 on Build GRCh38 Loss LOC100130451 SPAG16
nsv834531 Chr2:213250900 - 213437519 on Build GRCh38 Loss LOC100130451 SPAG16

View Full Product Details