Product Details

Assay Reference Genome
Location

Chr.1:6809398 on build GRCh38
Cytoband
1p36.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
23261
Gene Symbol
CAMTA1
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
CANPMR
Location
Chr.1:6785324-7769706 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CAMTA1 NM_001195563.1 NP_001182492.1
NM_001242701.1 NP_001229630.1
NM_015215.3 NP_056030.1
NR_038934.1
XM_011541083.2 XP_011539385.1
XM_011541084.2 XP_011539386.1
XM_011541085.2 XP_011539387.1
XM_011541086.2 XP_011539388.1
XM_011541087.2 XP_011539389.1
XM_011541088.2 XP_011539390.1
XM_011541089.2 XP_011539391.1
XM_011541090.2 XP_011539392.1
XM_011541091.2 XP_011539393.1
XM_011541092.2 XP_011539394.1
XM_017000773.1 XP_016856262.1
XM_017000774.1 XP_016856263.1
XM_017000775.1 XP_016856264.1
XM_017000776.1 XP_016856265.1
XM_017000777.1 XP_016856266.1
XM_017000778.1 XP_016856267.1
XM_017000779.1 XP_016856268.1
XM_017000780.1 XP_016856269.1
XM_017000781.1 XP_016856270.1
AB020640.3
AY037153.1 AAK67633.1
BC116457.1
BC151835.1
BG027385.1
CD103791.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1004598 Chr1:6645296 - 7178603 on Build GRCh38 Gain DNAJC11 CAMTA1 LOC100505887 LOC100129476

View Full Product Details