Product Details

Assay Reference Genome
Location

Chr.X:28597894 on build GRCh38
Cytoband
Xp21.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
11141
Gene Symbol
IL1RAPL1
Gene Name
interleukin 1 receptor accessory protein like 1
Gene Aliases
IL1R8, IL1RAPL, MRX10, MRX21, MRX34, OPHN4, TIGIRR-2
Location
Chr.X:28587564-29956350 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
IL1RAPL1 NM_014271.3 NP_055086.1
AF181284.1 AAF59411.1
AF284435.1 AAG21369.1
AJ243874.1 CAB56046.1
DN994486.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv6841 ChrX:28580318 - 28610714 on Build GRCh38 Insertion IL1RAPL1
nsv521056 ChrX:28590019 - 28664578 on Build GRCh38 Gain IL1RAPL1

View Full Product Details