Product Details
- Assay Reference Genome
Location
Chr.X:104572253 on build GRCh38
- Cytoband
- Xq22.3
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 26280
- Gene Symbol
- IL1RAPL2
- Gene Name
- interleukin 1 receptor accessory protein like 2
- Gene Aliases
- IL-1R9, IL1R9, IL1RAPL-2, TIGIRR-1
- Location
- Chr.X:104566315-105769729 on build GRCh38
- Assay Gene Location
- Within Intron 1
View Full Product Details