Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation ID |
Location |
CNV Subtype |
Genes |
nsv521056 |
ChrX:28590019 - 28664578 on Build GRCh38 |
Gain |
IL1RAPL1
|
Product Details
- Assay Reference Genome
Location
Chr.X:28611669 on build GRCh38
- Cytoband
- Xp21.3
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 11141
- Gene Symbol
- IL1RAPL1
- Gene Name
- interleukin 1 receptor accessory protein like 1
- Gene Aliases
- IL1R8, IL1RAPL, MRX10, MRX21, MRX34, OPHN4, TIGIRR-2
- Location
- Chr.X:28587564-29956350 on build GRCh38
- Assay Gene Location
- Within Intron 1
Target Copy Number Variation Details
- DGV Version
- Release date: 2016-05-15, GRCh GRCh38
Target Variation |
Location |
CNV Subtype |
Genes |
nsv521056 |
ChrX:28590019 - 28664578 on Build GRCh38 |
Gain |
IL1RAPL1
|
View Full Product Details