Product Details

Assay Reference Genome
Location

Chr.15:86080160 on build GRCh38
Cytoband
15q25.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
123624
Gene Symbol
AGBL1
Gene Name
ATP/GTP binding protein like 1
Gene Aliases
CCP4, FECD8
Location
Chr.15:86079635-87031476 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
AGBL1 XM_011521226.2 XP_011519528.1
XM_011521227.2 XP_011519529.1
XM_011521228.2 XP_011519530.1
XM_011521230.2 XP_011519532.1
XM_017021918.1 XP_016877407.1
XM_017021919.1 XP_016877408.1
XM_017021920.1 XP_016877409.1
XM_017021921.1 XP_016877410.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1648 Chr15:86078294 - 86119577 on Build GRCh38 Deletion LINC01584 AGBL1

View Full Product Details