Product Details

Assay Reference Genome
Location

Chr.10:75450585 on build GRCh38
Cytoband
10q22.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
83938
Gene Symbol
C10orf11
Gene Name
chromosome 10 open reading frame 11
Gene Aliases
CDA017
Location
Chr.10:75431646-76559220 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
C10orf11 NM_001305581.1 NP_001292510.1
XM_011540256.2 XP_011538558.1
XM_011540257.2 XP_011538559.1
XM_017016759.1 XP_016872248.1
XM_017016760.1 XP_016872249.1
XM_017016761.1 XP_016872250.1
BM720401.1
BX105111.1
HY006640.1

View Full Product Details