Product Details

Assay Reference Genome
Location

Chr.10:81902805 on build GRCh38
Cytoband
10q23.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
10718
Gene Symbol
NRG3
Gene Name
neuregulin 3
Gene Aliases
HRG3, pro-NRG3
Location
Chr.10:81875298-82989979 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NRG3 NM_001010848.3 NP_001010848.2
NM_001165972.1 NP_001159444.1
NM_001165973.1 NP_001159445.1
XM_005269444.4 XP_005269501.1
XM_011539172.2 XP_011537474.1
XM_011539173.2 XP_011537475.1
XM_011539175.2 XP_011537477.1
XM_017015573.1 XP_016871062.1
XM_017015574.1 XP_016871063.1
XM_017015575.1 XP_016871064.1
XM_017015576.1 XP_016871065.1
XM_017015577.1 XP_016871066.1
XM_017015578.1 XP_016871067.1
XM_017015579.1 XP_016871068.1
XM_017015580.1 XP_016871069.1
XM_017015581.1 XP_016871070.1
XM_017015584.1 XP_016871073.1
AK122886.1
AK296563.1
BC136811.1
DQ001411.1 AAY17216.1
DQ338459.1 ABC69293.1
DQ857894.1
HM068873.1
HM068874.1
HM068875.1
HM068876.1
HM068877.1
HM068878.1
HM068879.1
HM068880.1
HM068881.1
HM068882.1
HM068883.1
HM068884.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv551669 Chr10:81374678 - 82080786 on Build GRCh38 Gain NRG3
esv3891873 Chr10:81025608 - 82340753 on Build GRCh38 Gain NRG3 NRG3-AS1
nsv831925 Chr10:81771475 - 81938851 on Build GRCh38 Gain NRG3
nsv551670 Chr10:81545860 - 81972366 on Build GRCh38 Loss NRG3

View Full Product Details