Product Details

Assay Reference Genome
Location

Chr.6:123822397 on build GRCh38
Cytoband
6q22.31
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
154215
Gene Symbol
NKAIN2
Gene Name
Na+/K+ transporting ATPase interacting 2
Gene Aliases
FAM77B, NKAIP2, TCBA, TCBA1
Location
Chr.6:123803840-124825652 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
NKAIN2 NM_001040214.2 NP_001035304.1
NM_001300737.1 NP_001287666.1
NM_001300738.1 NP_001287667.1
NM_001300740.1 NP_001287669.1
NM_153355.4 NP_699186.2
XM_011535501.2 XP_011533803.1
XM_011535503.2 XP_011533805.1
XM_017010318.1 XP_016865807.1
XM_017010319.1 XP_016865808.1
AB070452.1 BAB85477.1
AK055326.1
AK308927.1
AK309385.1
AK309407.1
AK309802.1
AK310763.1
AK315886.1
BC035062.2 AAH35062.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv524559 Chr6:123189871 - 124008531 on Build GRCh38 Gain NKAIN2 TRDN HRAT13
nsv1018599 Chr6:123187871 - 124008918 on Build GRCh38 Gain NKAIN2 TRDN HRAT13

View Full Product Details