Product Details

Assay Reference Genome
Location

Chr.3:84970790 on build GRCh38
Cytoband
3p12.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
253559
Gene Symbol
CADM2
Gene Name
cell adhesion molecule 2
Gene Aliases
IGSF4D, NECL3, Necl-3, SynCAM 2, synCAM2
Location
Chr.3:84958982-86074429 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CADM2 NM_001167674.1 NP_001161146.1
NM_001167675.1 NP_001161147.1
XM_006713081.3 XP_006713144.1
XM_006713082.3 XP_006713145.1
XM_017006062.1 XP_016861551.1
AB646739.1
AK295885.1
AL834270.1 CAD38945.1
BC105999.1 AAI06000.1
BC112136.1 AAI12137.1
BC112138.1 AAI12139.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1010140 Chr3:84947861 - 84985130 on Build GRCh38 Gain CADM2
nsv1002058 Chr3:84707893 - 85340726 on Build GRCh38 Gain SNORA95 LINC00971 CADM2
nsv471620 Chr3:84831663 - 84982335 on Build GRCh38 Loss LINC00971 CADM2

View Full Product Details