Product Details

Assay Reference Genome
Location

Chr.2:79530802 on build GRCh38
Cytoband
2p12
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1496
Gene Symbol
CTNNA2
Gene Name
catenin alpha 2
Gene Aliases
CAP-R, CAPR, CT114, CTNR
Location
Chr.2:79512934-80648868 on build GRCh38
Assay Gene Location
Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CTNNA2 NM_001164883.1 NP_001158355.1
NM_001282597.2 NP_001269526.1
NM_001282598.1 NP_001269527.1
NM_004389.3 NP_004380.2
XM_011532556.2 XP_011530858.1
XM_017003403.1 XP_016858892.1
XM_017003404.1 XP_016858893.1
XM_017003405.1 XP_016858894.1
AK127226.1
AK295181.1
AK303035.1
AL708938.1
BC040458.1 AAH40458.1
BC052996.1 AAH52996.1
DA313393.1
DC341423.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv458407 Chr2:79523465 - 79584191 on Build GRCh38 Loss CTNNA2
esv3591408 Chr2:79524343 - 79533991 on Build GRCh38 Loss CTNNA2

View Full Product Details